Assay Details
| Gene Symbol: Slc37a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145960.1 | NP_001139432.1 | 17 - 18 | 1868 | 72 | |
| GenBank mRNA | AK028984.1 | - | 17 - 18 | 1544 | 72 | |
| AK220151.1 | - | 15 - 16 | 1310 | 72 | ||
| BC022752.1 | - | 3 - 4 | 243 | 72 | ||
| BC038000.1 | - | 17 - 18 | 1539 | 72 | ||
| BC048836.1 | - | 3 - 4 | 183 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc37a2
Genomic Map