Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900017064.mm.1 |
NM_001081429.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017064.mm.1 |
NM_001081429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017064.mm.1 |
TC0900002047.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017063.mm.1 |
NM_001081429.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017063.mm.1 |
NM_001081429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017063.mm.1 |
TC0900002047.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017066.mm.1 |
NM_001081429.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017066.mm.1 |
NM_001081429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017066.mm.1 |
TC0900002047.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900017065.mm.1 |
NM_001081429.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017065.mm.1 |
NM_001081429.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900017065.mm.1 |
TC0900002047.mm.1 |
MTA TC |