Assay Details
| Gene Symbol: Zbtb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033324.2 | NP_001028496.1 | 3 - 4 | 1611 | 67 | |
| GenBank mRNA | AK040211.1 | - | 3 - 4 | 1604 | 67 | |
| AK142947.1 | - | 3 - 4 | 1612 | 67 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb16
Genomic Map