Assay Details
| Gene Symbol: Zbtb16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033324.2 | NP_001028496.1 | 4 - 5 | 1699 | 64 | |
| GenBank mRNA | AK040211.1 | - | 4 - 5 | 1692 | 64 | |
| AK142947.1 | - | 4 - 5 | 1700 | 64 | ||
| BC132441.1 | - | 3 - 4 | 1387 | 64 | ||
| BC132443.1 | - | 3 - 4 | 1387 | 64 | ||
| BC138775.1 | - | 3 - 4 | 1510 | 8861797 | 64 | |
| Z47205.1 | - | 4 - 5 | 1740 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zbtb16
Genomic Map