Assay Details
| Gene Symbol: Nnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010924.2 | NP_035054.1 | 1 - 2 | 108 | ||
| GenBank mRNA | BC028757.1 | - | 1 - 2 | 191 | 1067421 | 108 |
| CF732455.1 | - | 1 - 2 | 108 | |||
| U86105.1 | - | 1 - 2 | 108 | |||
| U86106.1 | - | 1 - 2 | 108 | |||
| U86107.1 | - | 1 - 2 | 322 | 108 | ||
| U86108.1 | - | 1 - 2 | 322 | 108 | ||
| U89377.1 | - | 1 - 2 | 108 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Nnmt
Genomic Map