Assay Details
| Gene Symbol: Snupn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178374.3 | NP_848461.1 | 4 - 5 | 442 | 82 | |
| GenBank mRNA | AK002717.1 | - | 4 - 5 | 458 | 82 | |
| AK013151.1 | - | 4 - 5 | 452 | 82 | ||
| AK155315.1 | - | 4 - 5 | 459 | 82 | ||
| BC023317.1 | - | 422 | 82 | |||
| BC049638.1 | - | 4 - 5 | 442 | 6589249 | 82 | |
| BC061258.1 | - | 4 - 5 | 447 | 30251661 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Snupn
Genomic Map