Assay Details
| Gene Symbol: Scamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301634.1 | NP_001288563.1 | 2 - 3 | 299 | 59 | |
| NM_001301635.1 | NP_001288564.1 | 2 - 3 | 182 | 59 | ||
| NM_020270.3 | NP_064666.1 | 2 - 3 | 182 | 59 | ||
| GenBank mRNA | AF241833.1 | - | 2 - 3 | 268 | 59 | |
| BC018613.1 | - | 2 - 3 | 146 | 4511417 | 59 | |
| CJ131214.1 | - | 2 - 3 | 165 | 59 | ||
| CJ148246.1 | - | 2 - 3 | 303 | 59 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF241833.1, BC018613.1, NM_001301634.1, NM_001301635.1, NM_020270.3 transcripts.
Set Membership: |
|

Scamp5
Genomic Map