Assay Details
| Gene Symbol: Scamp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301634.1 | NP_001288563.1 | 4 - 5 | 584 | 59 | |
| NM_020270.3 | NP_064666.1 | 4 - 5 | 467 | 59 | ||
| GenBank mRNA | AF241833.1 | - | 4 - 5 | 553 | 59 | |
| BC018613.1 | - | 4 - 5 | 431 | 4511417 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scamp5
Genomic Map