Assay Details
| Gene Symbol: Zfp558 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028935.1 | NP_083211.1 | 6 - 7 | 745 | 101 | |
| GenBank mRNA | AK016584.1 | - | 8 - 9 | 2399 | 101 | |
| BC048749.1 | - | 5 - 6 | 453 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Zfp558
Genomic Map