Assay Details
| Gene Symbol: Lipe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039507.2 | NP_001034596.1 | 1 - 2 | 52 | 62 | |
| GenBank mRNA | AK169858.1 | - | 1 - 2 | 47 | 62 | |
| BC021642.1 | - | 1 - 2 | 48 | 4188567 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK169858.1, BC021642.1, NM_001039507.2 transcripts.
Set Membership: |
|

Lipe
Genomic Map