Assay Details
| Gene Symbol: Lipe | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039507.2 | NP_001034596.1 | 3 - 4 | 680 | 62 | |
| NM_010719.5 | NP_034849.2 | 3 - 4 | 1116 | 62 | ||
| GenBank mRNA | AK019846.1 | - | 3 - 4 | 1517 | 62 | |
| AK029984.1 | - | 3 - 4 | 1119 | 62 | ||
| AK169858.1 | - | 3 - 4 | 675 | 62 | ||
| BC021642.1 | - | 3 - 4 | 676 | 4188567 | 62 | |
| BC053502.1 | - | 3 - 4 | 740 | 62 | ||
| U08188.1 | - | 2 - 3 | 1201 | 62 | ||
More Information
Set Membership: |
|

Lipe
Genomic Map