Assay Details
| Gene Symbol: Rabac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010261.2 | NP_034391.1 | 1 - 2 | 203 | 79 | |
| GenBank mRNA | AF120162.1 | - | 1 - 2 | 147 | 79 | |
| AF252856.1 | - | 1 - 2 | 137 | 79 | ||
| AK008559.1 | - | 1 - 2 | 115 | 79 | ||
| AK009726.1 | - | 1 - 2 | 63 | 79 | ||
| AK150681.1 | - | 1 - 2 | 84 | 79 | ||
| AK170289.1 | - | 1 - 2 | 163 | 79 | ||
| BC008242.1 | - | 1 - 2 | 136 | 2655184 | 79 | |
| BY127932.1 | - | 1 - 2 | 204 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rabac1
Genomic Map