Assay Details
Gene Symbol: Syde1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027875.1 | NP_082151.1 | 1 - 2 | 164 | 100 | |
GenBank mRNA | AK004654.1 | - | 1 - 2 | 146 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
