Assay Details
| Gene Symbol: Syde1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027875.1 | NP_082151.1 | 4 - 5 | 1366 | 80 | |
| GenBank mRNA | AK004654.1 | - | 4 - 5 | 1348 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Syde1
Genomic Map