Assay Details
| Gene Symbol: Dnmt3l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081695.2 | NP_001075164.1 | 4 - 5 | 493 | 80 | |
| NM_001284197.1 | NP_001271126.1 | 2 - 3 | 334 | 80 | ||
| NM_019448.4 | NP_062321.1 | 3 - 4 | 347 | 80 | ||
| GenBank mRNA | AF220524.1 | - | 2 - 3 | 278 | 80 | |
| AJ404467.1 | - | 2 - 3 | 319 | 80 | ||
| AK010434.1 | - | 3 - 4 | 348 | 80 | ||
| AV473569.1 | - | 2 - 3 | 334 | 80 | ||
| BC083147.1 | - | 2 - 3 | 332 | 30447906 | 80 | |
| EF051624.1 | - | 4 - 5 | 493 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dnmt3l
Genomic Map