Assay Details
| Gene Symbol: Casp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009809.5 | NP_033939.1 | 5 - 6 | 780 | 70 | |
| GenBank mRNA | AF092997.1 | - | 4 - 5 | 625 | 70 | |
| AJ007750.1 | - | 5 - 6 | 645 | 70 | ||
| AK081645.1 | - | 5 - 6 | 733 | 70 | ||
| BC101959.1 | - | 5 - 6 | 718 | 40044873 | 70 | |
| BC101960.1 | - | 5 - 6 | 718 | 40044874 | 70 | |
| BC103555.1 | - | 5 - 6 | 718 | 40044870 | 70 | |
| BC103556.1 | - | 5 - 6 | 718 | 40044871 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Casp14
Genomic Map