Assay Details
| Gene Symbol: Ldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252658.1 | NP_001239587.1 | 12 - 13 | 2039 | 69 | |
| NM_001252659.1 | NP_001239588.1 | 12 - 13 | 2036 | 69 | ||
| NM_010700.3 | NP_034830.2 | 12 - 13 | 2039 | 69 | ||
| GenBank mRNA | AF425607.1 | - | 12 - 13 | 1887 | 69 | |
| AK149950.1 | - | 12 - 13 | 1977 | 69 | ||
| AK151968.1 | - | 12 - 13 | 2006 | 69 | ||
| AK152103.1 | - | 12 - 13 | 1975 | 69 | ||
| AK160006.1 | - | 12 - 13 | 1977 | 69 | ||
| AK166466.1 | - | 12 - 13 | 1976 | 69 | ||
| AK170264.1 | - | 12 - 13 | 1984 | 69 | ||
| BC019207.1 | - | 12 - 13 | 1957 | 5052599 | 69 | |
| BC053041.1 | - | 12 - 13 | 1984 | 5704871 | 69 | |
| X64414.1 | - | 12 - 13 | 1958 | 69 | ||
| Z19521.1 | - | 12 - 13 | 1837 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ldlr
Genomic Map