Assay Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104616.1 | NP_001098086.1 | 10 - 11 | 1232 | 86 | |
| NM_009041.3 | NP_033067.2 | 10 - 11 | 1638 | 86 | ||
| GenBank mRNA | AK146423.1 | - | 10 - 11 | 1296 | 86 | |
| AK156370.1 | - | 10 - 11 | 1324 | 86 | ||
| AK162204.1 | - | 10 - 11 | 1210 | 86 | ||
| AK168449.1 | - | 10 - 11 | 1309 | 86 | ||
| BC034438.1 | - | 10 - 11 | 1464 | 86 | ||
| X60672.1 | - | 10 - 11 | 1251 | 86 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rdx
Genomic Map