Assay Details
| Gene Symbol: Rdx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104616.1 | NP_001098086.1 | 11 - 12 | 1400 | 59 | |
| NM_009041.3 | NP_033067.2 | 11 - 12 | 1806 | 59 | ||
| GenBank mRNA | AK146423.1 | - | 11 - 12 | 1464 | 59 | |
| AK156370.1 | - | 11 - 12 | 1492 | 59 | ||
| AK162204.1 | - | 11 - 12 | 1378 | 59 | ||
| AK162224.1 | - | 1 - 2 | 70 | 59 | ||
| AK168449.1 | - | 11 - 12 | 1477 | 59 | ||
| AK217944.1 | - | 1 - 2 | 75 | 59 | ||
| BC034438.1 | - | 11 - 12 | 1632 | 59 | ||
| X60672.1 | - | 11 - 12 | 1419 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rdx
Genomic Map