Assay Details
Gene Symbol: 3110039I08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK014147.1 | - | 405 | 103 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome |
---|

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 666639 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 145336 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 544137 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 648330 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 791937 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265431 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506418 | AK005789.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506418 | BC048623.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 506418 | NM_029297.1 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 266690 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639713 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 264733 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 53634 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 687931 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 833262 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396911 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 26847 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 736580 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 522893 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6199 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 135287 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 718270 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 179108 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 92612 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 154856 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01912742 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01912778 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06761419 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06761508 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06761457 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P01912778 | - | - |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06761457 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448703 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448721 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448702 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08155901 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448691 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448858 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448674 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448694 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448689 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448686 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448697 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08040139 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448666 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448699 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08114181 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448844 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448838 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08040133 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448719 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448693 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448839 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07501692 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448860 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448841 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448705 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07941505 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448848 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448849 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448851 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448718 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08096587 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448680 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07588172 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448663 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448711 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448714 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448715 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448665 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448687 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448712 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07687581 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448659 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448828 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448690 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448685 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448832 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448684 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07764865 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448670 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07941507 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448683 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448695 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448667 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07494435 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448829 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448701 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448830 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07941515 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448836 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26448831 | - | - |
