Assay Details
| Gene Symbol: Ddx10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029936.2 | NP_084212.2 | 10 - 11 | 1435 | 142 | |
| GenBank mRNA | AK019495.1 | - | 10 - 11 | 1415 | 142 | |
| AK052540.1 | - | 10 - 11 | 1415 | 142 | ||
| BC023303.1 | - | 10 - 11 | 1383 | 142 | ||
| BC049261.1 | - | 10 - 11 | 1376 | 142 | ||
| BC055481.1 | - | 9 - 10 | 1153 | 142 | ||
| BC080729.1 | - | 10 - 11 | 1403 | 6504816 | 142 | |
More Information
Set Membership: |
|

Ddx10
Genomic Map