Assay Details
| Gene Symbol: Ddx10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029936.2 | NP_084212.2 | 4 - 5 | 641 | 111 | |
| GenBank mRNA | AK019495.1 | - | 4 - 5 | 621 | 111 | |
| AK045032.1 | - | 4 - 5 | 606 | 111 | ||
| AK052540.1 | - | 4 - 5 | 621 | 111 | ||
| BC023303.1 | - | 4 - 5 | 589 | 111 | ||
| BC049261.1 | - | 4 - 5 | 582 | 111 | ||
| BC055481.1 | - | 3 - 4 | 359 | 111 | ||
| BC080729.1 | - | 4 - 5 | 609 | 6504816 | 111 | |
More Information
Set Membership: |
|

Ddx10
Genomic Map