Assay Details
| Gene Symbol: Kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212445.2 | NP_997610.1 | 4 - 5 | 966 | 88 | |
| GenBank mRNA | AK075811.1 | - | 4 - 5 | 775 | 88 | |
More Information
Set Membership: |
|

Kdelc2
Genomic Map