Assay Details
| Gene Symbol: Specc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145826.1 | NP_001139298.1 | 15 - 16 | 3401 | 84 | |
| NM_153406.3 | NP_700455.3 | 12 - 13 | 3172 | 84 | ||
| GenBank mRNA | AB093236.1 | - | 15 - 16 | 3401 | 84 | |
| AY884297.1 | - | 12 - 13 | 3173 | 84 | ||
| BC037464.1 | - | 9 - 10 | 2259 | 4194868 | 84 | |
| BC072595.1 | - | 11 - 12 | 3011 | 6408832 | 84 | |
More Information
Updated Mapping Information
Set Membership: |
|

Specc1l
Genomic Map