Assay Details
| Gene Symbol: Specc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145826.1 | NP_001139298.1 | 16 - 17 | 3496 | 69 | |
| NM_153406.3 | NP_700455.3 | 13 - 14 | 3267 | 69 | ||
| GenBank mRNA | AB093236.1 | - | 16 - 17 | 3496 | 69 | |
| AK015541.1 | - | 2 - 3 | 232 | 69 | ||
| AK016555.1 | - | 2 - 3 | 233 | 69 | ||
| AY884297.1 | - | 13 - 14 | 3268 | 69 | ||
| BC037464.1 | - | 10 - 11 | 2354 | 4194868 | 69 | |
| BC072595.1 | - | 12 - 13 | 3106 | 6408832 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Specc1l
Genomic Map