Assay Details
| Gene Symbol: Specc1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145826.1 | NP_001139298.1 | 8 - 9 | 2561 | 80 | |
| NM_153406.3 | NP_700455.3 | 5 - 6 | 2332 | 80 | ||
| GenBank mRNA | AB093236.1 | - | 8 - 9 | 2561 | 80 | |
| AK041543.1 | - | 2 - 3 | 825 | 80 | ||
| AY884297.1 | - | 5 - 6 | 2333 | 80 | ||
| BC037464.1 | - | 2 - 3 | 1419 | 4194868 | 80 | |
| BC072595.1 | - | 4 - 5 | 2171 | 6408832 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Specc1l
Genomic Map