Assay Details
| Gene Symbol: Gnaz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010311.3 | NP_034441.1 | 1 - 2 | 154 | 59 | |
| GenBank mRNA | AK080716.1 | - | 1 - 2 | 154 | 59 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK080716.1, NM_010311.3 transcripts.
Set Membership: |
|

Gnaz
Genomic Map