Assay Details
| Gene Symbol: Tmprss9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081688.2 | NP_001075157.2 | 8 - 9 | 1122 | 86 | |
| GenBank mRNA | AB109391.1 | - | 8 - 9 | 1233 | 86 | |
| BC158102.1 | - | 8 - 9 | 1218 | 9007387 | 86 | |
| BC172169.1 | - | 8 - 9 | 1218 | 9054839 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss9
Genomic Map