Assay Details
| Gene Symbol: Tmprss9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081688.2 | NP_001075157.2 | 13 - 14 | 2164 | 90 | |
| GenBank mRNA | BC158102.1 | - | 13 - 14 | 2260 | 9007387 | 90 |
| BC172169.1 | - | 13 - 14 | 2260 | 9054839 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tmprss9
Genomic Map