Assay Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145400.3 | NP_663375.3 | 10 - 11 | 1704 | 101 | |
| GenBank mRNA | AK028721.1 | - | 10 - 11 | 1680 | 101 | |
| AK172893.1 | - | 10 - 11 | 1664 | 101 | ||
| BC006649.1 | - | 11 - 12 | 1843 | 3498834 | 101 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube4a
Genomic Map