Assay Details
| Gene Symbol: Ube4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145400.3 | NP_663375.3 | 13 - 14 | 2258 | 100 | |
| GenBank mRNA | AK028721.1 | - | 12 - 13 | 1929 | 100 | |
| AK172893.1 | - | 13 - 14 | 2218 | 100 | ||
| BC006649.1 | - | 14 - 15 | 2397 | 3498834 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube4a
Genomic Map