Assay Details
| Gene Symbol: Anxa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007585.3 | NP_031611.1 | 1 - 2 | 60 | 71 | |
| GenBank mRNA | AK012563.1 | - | 1 - 2 | 62 | 71 | |
| AK088645.1 | - | 1 - 2 | 49 | 71 | ||
| AK146096.1 | - | 1 - 2 | 48 | 71 | ||
| AK150586.1 | - | 1 - 2 | 48 | 71 | ||
| AK152561.1 | - | 1 - 2 | 48 | 71 | ||
| AK160746.1 | - | 1 - 2 | 48 | 71 | ||
| AK164507.1 | - | 1 - 2 | 53 | 71 | ||
| AK165888.1 | - | 1 - 2 | 48 | 71 | ||
| AK167894.1 | - | 1 - 2 | 47 | 71 | ||
| AK168083.1 | - | 1 - 2 | 48 | 71 | ||
| BC003327.1 | - | 1 - 2 | 60 | 3156634 | 71 | |
| BC005763.1 | - | 1 - 2 | 34 | 2655513 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK012563.1, AK088645.1, AK146096.1, AK150586.1, AK152561.1, AK164507.1, AK165888.1, AK167894.1, AK168083.1, BC003327.1, BC005763.1, NM_007585.3 transcripts.
Set Membership: |
|

Anxa2
Genomic Map