Assay Details
Gene Symbol: Olfr894 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146868.2 | NP_667079.1 | 1 | 479 | 115 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P369330 | NM_146868.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P369330 | NM_146868.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P369330 | NM_146868.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P369330 | NM_146868.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369330 | NM_146868.2 | mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P369330 | NM_146868.2 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434003 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434036 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434024 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434016 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434020 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434035 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433994 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434055 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434031 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434033 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434021 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434057 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434054 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433997 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434026 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434017 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434052 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434043 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434048 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434051 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434032 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434049 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434058 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434034 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26433995 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434015 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434019 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434027 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434000 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26434056 | - | - |
Whole Mouse Genome G4122A | A_51_P369330 | NM_146868.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P369330 | NM_146868.2 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P121937 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P107880 | - | - |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117316 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434026 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433995 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434019 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434060 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433989 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434034 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434036 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434003 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434047 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434005 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434024 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434009 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434007 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434017 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434027 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433990 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434006 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434020 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434058 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434014 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434000 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433994 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433986 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434041 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433993 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26433985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26434016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07587808 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434016 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434035 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434006 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434000 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434021 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434019 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434050 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434047 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26433994 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434026 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434055 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434041 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434033 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26433997 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434051 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434034 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434011 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434027 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434007 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434031 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08080117 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434022 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434054 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434010 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434003 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434020 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434009 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434004 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434049 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434060 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07735669 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434017 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07755501 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434056 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07561272 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434057 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26433995 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434015 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434014 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434032 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434005 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434043 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07735676 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26434058 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07515584 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10584385 | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10584384 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 465568 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 2760601 | - | - |
Illumina mouse-6 v1.0 expression beadchip | 2760601 | - | - |
Illumina mouseRef-8 v1.0 expression beadchip | 0002760601 | - | - |
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1228252 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1228252 | - | - |
mouse-6 v1.1 (Illumina) | scl37102.1.1_24-S | - | - |
Illumina Sentrix mouse-6.v1 bead array | 2760601 | - | - |
Illumina Sentrix MouseRef-8 Expression BeadChip | scl37102.1.1_24-S | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2760601 | - | - |
MouseRef-8 v2 Expression BeadChip | ILMN_2631775 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2631775 | - | - |
