Assay Details
Gene Symbol: Gldnos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045805.1 | 1 - 2 | 135 | 102 | ||
GenBank mRNA | AK148218.1 | - | 1 - 2 | 136 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
