Assay Details
Gene Symbol: Gldnos | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045805.1 | 2 - 3 | 177 | 63 | ||
GenBank mRNA | AK148218.1 | - | 2 - 3 | 178 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P117152 | AK148218.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P117152 | AK148218.1 | mouse_transcriptome |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07832466 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07835256 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07832467 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524952 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524945 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524955 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524896 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524876 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524947 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524911 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524927 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07732186 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524956 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524925 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524914 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524886 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524940 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524890 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07922651 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524894 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524909 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08121905 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524907 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524883 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524957 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524917 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07734504 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07555878 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524916 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524891 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524963 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524881 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524922 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524942 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524878 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524939 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07611688 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524946 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524930 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524882 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524948 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524949 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524962 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524898 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524959 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07867818 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524903 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524900 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P07858845 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524935 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524920 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524937 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524933 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524877 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524910 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P08121909 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524884 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524943 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524931 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524928 | - | - |
Agilent Six_ChIP_02_of_03_v2 | A_68_P26524913 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0900019389.mm.1 | NR_045805.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900019389.mm.1 | NR_045805.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0900019389.mm.1 | TC0900002319.mm.1 | MTA TC |
Murine Genome U74v2 Set | 169082_PM_at | - | - |
Murine Genome U74v2 Set | 169082_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 369160 | - | - |
ABI Mouse Genome Survey Microarray | 521841 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 103830091 | - | - |
