Assay Details
| Gene Symbol: Panx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172454.2 | NP_766042.2 | 1 - 2 | 352 | 61 | |
| GenBank mRNA | AK028288.1 | - | 1 - 2 | 319 | 61 | |
| AK036933.1 | - | 1 - 2 | 308 | 61 | ||
| BC052822.1 | - | 1 - 2 | 320 | 30058169 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Panx3
Genomic Map