Assay Details
| Gene Symbol: Panx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172454.2 | NP_766042.2 | 2 - 3 | 493 | 66 | |
| GenBank mRNA | AK028288.1 | - | 2 - 3 | 460 | 66 | |
| AK036933.1 | - | 2 - 3 | 449 | 66 | ||
| BC052822.1 | - | 2 - 3 | 461 | 30058169 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Panx3
Genomic Map