Assay Details
| Gene Symbol: Dpy19l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166207.1 | NP_001159679.1 | 19 - 20 | 2074 | 68 | |
| GenBank mRNA | AK029497.1 | - | 19 - 20 | 2078 | 68 | |
| AK030128.1 | - | 19 - 20 | 2063 | 68 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatogenesis
spermatid development
protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
cell differentiation
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity
multicellular organism development
spermatogenesis
spermatid development
protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
cell differentiation
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2235136 | AK029497.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2235136 | AV254516.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2235136 | NM_001166207.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 387972 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 458485 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 783865 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 450259 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 114850 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 289629 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498992 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 712778 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 525982 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901478 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154013 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 325404 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 543364 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285374 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 750005 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 830518 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 717664 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 605213 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 723997 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 609356 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 142388 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 36380 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 61116 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 192369 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 500853 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 112848 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 254964 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 592757 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 838112 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 498065 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 586534 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 747737 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 680632 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 455889 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 235841 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 814375 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 576159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 441159 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 275830 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 373541 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 654753 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 22053 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 714917 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 257958 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 304464 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 402863 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 83317 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 258614 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 662376 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 919597 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 355613 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 793702 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 227839 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 869308 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 45622 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 13135 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 73110 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 35552 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 115004 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 38866 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 163146 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119921 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 39194 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 16482 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 23067 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 128851 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 75041 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P122622 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P115609 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2235136 | AK029497.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2235136 | AV254516.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2235136 | NM_001166207.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725737 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725778 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725712 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725717 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P122622 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725684 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P115609 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725692 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725662 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725786 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06725683 | - | - |
| Whole Mouse Genome G4122A | A_51_P211770 | AK030128.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P211770 | AK029497.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P211770 | NM_001166207.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P126786 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06725662 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P115609 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06725745 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P122622 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06725724 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06725684 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08030189 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363031 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08031914 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362988 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363041 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08031920 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08090161 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362989 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363007 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363021 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362982 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363036 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363038 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363017 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362997 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363026 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363008 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08123103 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07911686 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363039 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362985 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363014 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362990 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363001 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362979 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362994 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363004 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363030 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363010 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362992 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362980 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363011 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363033 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363034 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26362981 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363012 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363027 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363023 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363024 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363018 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26363029 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10591851 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591852 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591847 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591848 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591850 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591849 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10591846 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1437272_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10591844 | - | - |
| Mouse Genome 430 2.0 Array | 1437272_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1437272_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900016170.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016170.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016170.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016164.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016164.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016164.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016162.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016162.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016162.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016163.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016163.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016163.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016166.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016166.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016166.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016167.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016167.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016167.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016169.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016169.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016169.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016161.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016161.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016161.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016171.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016171.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016171.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016165.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016165.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016165.mm.1 | TC0900001923.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900016168.mm.1 | NM_001166207.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016168.mm.1 | NM_001166207.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900016168.mm.1 | TC0900001923.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1437272_PM_at | - | - |
| Mouse Expression Set 430 | 1437272_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437272_PM_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1437272_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 874720 | - | - |

Dpy19l2
Genomic Map