Assay Details
| Gene Symbol: Nrgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022029.2 | NP_071312.1 | 2 - 3 | 432 | 145 | |
| GenBank mRNA | AK002933.1 | - | 2 - 3 | 392 | 145 | |
| AK158630.1 | - | 2 - 3 | 419 | 145 | ||
| BC061102.1 | - | 2 - 3 | 396 | 6590375 | 145 | |
| BC138511.1 | - | 2 - 3 | 309 | 8861531 | 145 | |
| BC138513.1 | - | 2 - 3 | 309 | 8861533 | 145 | |
| BU604757.1 | - | 2 - 3 | 435 | 145 | ||
More Information
Set Membership: |
|

Nrgn
Genomic Map