Assay Details
| Gene Symbol: Cyp19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007810.3 | NP_031836.1 | 1 - 2 | 192 | 83 | |
| GenBank mRNA | AK143323.1 | - | 1 - 2 | 272 | 83 | |
| BC103670.1 | - | 1 - 2 | 175 | 40047093 | 83 | |
| BC104489.1 | - | 1 - 2 | 175 | 40047095 | 83 | |
| D00659.1 | - | 1 - 2 | 197 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp19a1
Genomic Map