Assay Details
| Gene Symbol: Cyp19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007810.3 | NP_031836.1 | 5 - 6 | 786 | 101 | |
| GenBank mRNA | AK143323.1 | - | 5 - 6 | 866 | 101 | |
| BC103670.1 | - | 5 - 6 | 769 | 40047093 | 101 | |
| BC104489.1 | - | 5 - 6 | 769 | 40047095 | 101 | |
| D00659.1 | - | 5 - 6 | 791 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp19a1
Genomic Map