Assay Details
| Gene Symbol: Cyp19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007810.3 | NP_031836.1 | 7 - 8 | 1066 | 110 | |
| GenBank mRNA | AK143323.1 | - | 7 - 8 | 1146 | 110 | |
| BC103670.1 | - | 7 - 8 | 1049 | 40047093 | 110 | |
| BC104489.1 | - | 7 - 8 | 1049 | 40047095 | 110 | |
| D00659.1 | - | 7 - 8 | 1071 | 110 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyp19a1
Genomic Map