Assay Details
| Gene Symbol: Ddx25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013932.4 | NP_038960.2 | 2 - 3 | 201 | 113 | |
| GenBank mRNA | AK050693.1 | - | 2 - 3 | 196 | 113 | |
| AK078340.1 | - | 2 - 3 | 160 | 113 | ||
| BC024852.1 | - | 2 - 3 | 145 | 5358404 | 113 | |
| BC061130.1 | - | 2 - 3 | 156 | 6774579 | 113 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK050693.1, AK078340.1, BC024852.1, BC061130.1 transcripts.
Set Membership: |
|

Ddx25
Genomic Map