| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_001190911.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_001190911.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_001190912.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_001190912.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_001190913.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_001190913.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_001190914.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_001190914.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_026324.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
NM_026324.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002346.mm.1 |
TC0900000313.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_001190911.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_001190911.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_001190912.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_001190912.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_001190913.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_001190913.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_001190914.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_001190914.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_026324.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
NM_026324.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900002347.mm.1 |
TC0900000313.mm.1 |
MTA TC |