Assay Details
| Gene Symbol: Car12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052639.1 | - | 8 - 9 | 998 | 69 | |
| AK170859.1 | - | 8 - 9 | 957 | 69 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Car12| Gene Symbol: Car12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK052639.1 | - | 8 - 9 | 998 | 69 | |
| AK170859.1 | - | 8 - 9 | 957 | 69 | ||
Set Membership: |
|