Assay Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008959.3 | NP_032985.2 | 11 - 12 | 1920 | 71 | |
| GenBank mRNA | AF042731.1 | - | 11 - 12 | 1335 | 71 | |
| AK036990.1 | - | 11 - 12 | 1382 | 71 | ||
| AK054101.1 | - | 11 - 12 | 1359 | 71 | ||
| AK085857.1 | - | 11 - 12 | 1921 | 71 | ||
| AK086136.1 | - | 11 - 12 | 1579 | 71 | ||
| AK088048.1 | - | 11 - 12 | 1402 | 71 | ||
| AK137681.1 | - | 11 - 12 | 1590 | 71 | ||
| AK172873.1 | - | 10 - 11 | 3706 | 71 | ||
| BC003260.1 | - | 11 - 12 | 1557 | 3501141 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptdss1
Genomic Map