Assay Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008959.3 | NP_032985.2 | 2 - 3 | 952 | 67 | |
| GenBank mRNA | AF042731.1 | - | 2 - 3 | 367 | 67 | |
| AK036990.1 | - | 2 - 3 | 414 | 67 | ||
| AK054101.1 | - | 2 - 3 | 391 | 67 | ||
| AK085857.1 | - | 2 - 3 | 953 | 67 | ||
| AK088048.1 | - | 2 - 3 | 434 | 67 | ||
| AK137681.1 | - | 2 - 3 | 622 | 67 | ||
| AK172873.1 | - | 2 - 3 | 929 | 67 | ||
| BC003260.1 | - | 2 - 3 | 589 | 3501141 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptdss1
Genomic Map