Assay Details
| Gene Symbol: Ptdss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008959.3 | NP_032985.2 | 5 - 6 | 1273 | 65 | |
| GenBank mRNA | AF042731.1 | - | 5 - 6 | 688 | 65 | |
| AK036990.1 | - | 5 - 6 | 735 | 65 | ||
| AK054101.1 | - | 5 - 6 | 712 | 65 | ||
| AK085857.1 | - | 5 - 6 | 1274 | 65 | ||
| AK088048.1 | - | 5 - 6 | 755 | 65 | ||
| AK137681.1 | - | 5 - 6 | 943 | 65 | ||
| AK172873.1 | - | 5 - 6 | 1250 | 65 | ||
| BC003260.1 | - | 5 - 6 | 910 | 3501141 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptdss1
Genomic Map