Assay Details
| Gene Symbol: Zfp26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK029298.1 | - | 4 - 5 | 1040 | 71 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK029298.1 transcripts.
Set Membership: |
|

Zfp26
Genomic Map