Assay Details
| Gene Symbol: Rinl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177158.5 | NP_796132.2 | 2 - 3 | 124 | 60 | |
| GenBank mRNA | AK030993.1 | - | 2 - 3 | 117 | 60 | |
| BC044761.1 | - | 2 - 3 | 160 | 60 | ||
| BC046530.1 | - | 2 - 3 | 160 | 5329601 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rinl
Genomic Map